Assay Details
| Gene Symbol: LOC101715129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252416.1 | XP_021108075.1 | 1 | 1117 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715129| Gene Symbol: LOC101715129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252416.1 | XP_021108075.1 | 1 | 1117 | 75 | |
Set Membership: |
|