Assay Details
| Gene Symbol: Gpr132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836838.3 | XP_004836895.1 | 5 - 6 | 1072 | 102 | |
| XM_021243445.1 | XP_021099104.1 | 8 - 9 | 2566 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836838.3, XM_021243445.1 transcripts.
Set Membership: |
|

Gpr132
Genomic Map