Assay Details
| Gene Symbol: LOC106007587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864168.3 | XP_004864225.1 | 3 - 4 | 625 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106007587| Gene Symbol: LOC106007587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864168.3 | XP_004864225.1 | 3 - 4 | 625 | 80 | |
Set Membership: |
|