Assay Details
| Gene Symbol: LOC101696361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866539.2 | XP_004866596.1 | 1 - 2 | 469 | 52 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696361| Gene Symbol: LOC101696361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866539.2 | XP_004866596.1 | 1 - 2 | 469 | 52 | |
Set Membership: |
|