Assay Details
| Gene Symbol: LOC101696361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866539.2 | XP_004866596.1 | 2 - 3 | 589 | 74 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696361| Gene Symbol: LOC101696361 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866539.2 | XP_004866596.1 | 2 - 3 | 589 | 74 | |