Assay Details
| Gene Symbol: Shmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868676.3 | XP_004868733.1 | 6 - 7 | 936 | 62 | |
| XM_004868677.3 | XP_004868734.1 | 7 - 8 | 1012 | 62 | ||
| XM_004868678.3 | XP_004868735.1 | 7 - 8 | 1008 | 62 | ||
More Information
Set Membership: |
|

Shmt1
Genomic Map