Assay Details
Gene Symbol: Slco6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013071176.2 | XP_012926630.1 | 4 - 5 | 1031 | 111 | |
XM_021255820.1 | XP_021111479.1 | 4 - 5 | 1031 | 111 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
