Assay Details
| Gene Symbol: LOC101725596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239331.1 | XP_021094990.1 | 7 - 8 | 777 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101725596| Gene Symbol: LOC101725596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239331.1 | XP_021094990.1 | 7 - 8 | 777 | 98 | |
Set Membership: |
|