Assay Details
| Gene Symbol: Bend2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245373.1 | XP_021101032.1 | 4 - 5 | 515 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bend2| Gene Symbol: Bend2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021245373.1 | XP_021101032.1 | 4 - 5 | 515 | 78 | |