Assay Details
Gene Symbol: Fam35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021246737.1 | XP_021102396.1 | 9 - 10 | 2975 | 90 | |
XM_021246738.1 | XP_021102397.1 | 9 - 10 | 2970 | 90 | ||
XM_021246739.1 | XP_021102398.1 | 8 - 9 | 2527 | 90 | ||
XM_021246740.1 | XP_021102399.1 | 8 - 9 | 2911 | 90 | ||
XM_021246742.1 | XP_021102401.1 | 7 - 8 | 2799 | 90 | ||
XM_021246743.1 | XP_021102402.1 | 7 - 8 | 2385 | 90 | ||
XM_021246744.1 | XP_021102403.1 | 7 - 8 | 3011 | 90 | ||
XM_021246746.1 | XP_021102405.1 | 9 - 10 | 2973 | 90 | ||
XM_021246749.1 | XP_021102408.1 | 8 - 9 | 1321 | 90 | ||
XM_021246750.1 | XP_021102409.1 | 8 - 9 | 1316 | 90 | ||
XM_021246751.1 | XP_021102410.1 | 7 - 8 | 860 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
