Assay Details
| Gene Symbol: Fam131a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834868.3 | XP_004834925.1 | 3 - 4 | 567 | 60 | |
| XM_013072313.2 | XP_012927767.1 | 2 - 3 | 200 | 60 | ||
| XM_021256135.1 | XP_021111794.1 | 4 - 5 | 475 | 60 | ||
More Information
Set Membership: |
|

Fam131a
Genomic Map