Assay Details
| Gene Symbol: Meltf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835180.2 | XP_004835237.1 | 10 - 11 | 2460 | 57 | |
| XM_021255204.1 | XP_021110863.1 | 10 - 11 | 2460 | 57 | ||
| XM_021255208.1 | XP_021110867.1 | 11 - 12 | 2079 | 57 | ||
| XM_021255213.1 | XP_021110872.1 | 11 - 12 | 2090 | 57 | ||
More Information
Set Membership: |
|

Meltf
Genomic Map