Assay Details
| Gene Symbol: Pitx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836217.2 | XP_004836274.1 | 2 - 3 | 274 | 85 | |
| XM_021238588.1 | XP_021094247.1 | 2 - 3 | 426 | 85 | ||
More Information
Set Membership: |
|

Pitx1
Genomic Map