Assay Details
| Gene Symbol: Adrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838882.2 | XP_004838939.1 | 1 - 2 | 3413 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adrb1| Gene Symbol: Adrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838882.2 | XP_004838939.1 | 1 - 2 | 3413 | 80 | |
Set Membership: |
|