Assay Details
| Gene Symbol: Adrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838882.2 | XP_004838939.1 | 2 - 3 | 4330 | 100 | |
| XM_021253410.1 | XP_021109069.1 | 3 - 4 | 4413 | 100 | ||
More Information
Set Membership: |
|

Adrb1
Genomic Map