Assay Details
| Gene Symbol: Cdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843111.3 | XP_004843168.1 | 8 - 9 | 1473 | 65 | |
| XM_004843112.2 | XP_004843169.1 | 8 - 9 | 1485 | 65 | ||
| XM_021256949.1 | XP_021112608.1 | 8 - 9 | 1487 | 65 | ||
| XM_021256950.1 | XP_021112609.1 | 8 - 9 | 1488 | 65 | ||
More Information
Set Membership: |
|

Cdh5
Genomic Map