Assay Details
| Gene Symbol: Erp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845918.2 | XP_004845975.1 | 5 - 6 | 730 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erp27| Gene Symbol: Erp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845918.2 | XP_004845975.1 | 5 - 6 | 730 | 56 | |
Set Membership: |
|