Assay Details
| Gene Symbol: Znf184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847808.3 | XP_004847865.1 | 2 - 3 | 519 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znf184| Gene Symbol: Znf184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847808.3 | XP_004847865.1 | 2 - 3 | 519 | 58 | |
Set Membership: |
|