Assay Details
| Gene Symbol: Znf184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847808.3 | XP_004847865.1 | 5 - 6 | 868 | 99 | |
| XM_021260777.1 | XP_021116436.1 | 4 - 5 | 743 | 99 | ||
More Information
Set Membership: |
|

Znf184
Genomic Map