Assay Details
| Gene Symbol: Ranbp3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848609.3 | XP_004848666.1 | 12 - 13 | 1566 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ranbp3l| Gene Symbol: Ranbp3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848609.3 | XP_004848666.1 | 12 - 13 | 1566 | 73 | |
Set Membership: |
|