Assay Details
| Gene Symbol: CUNH17orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857347.2 | XP_004857404.1 | 2 - 3 | 256 | 70 | |
| XM_004857348.2 | XP_004857405.1 | 2 - 3 | 256 | 70 | ||
| XM_004857350.2 | XP_004857407.1 | 2 - 3 | 256 | 70 | ||
| XM_021238186.1 | XP_021093845.1 | 2 - 3 | 256 | 70 | ||
More Information
Set Membership: |
|

CUNH17orf49
Genomic Map