Assay Details
| Gene Symbol: Map3k20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857686.3 | XP_004857743.1 | 17 - 18 | 1668 | 74 | |
| XM_004857687.3 | XP_004857744.1 | 17 - 18 | 1797 | 74 | ||
| XM_004857688.2 | XP_004857745.1 | 17 - 18 | 1637 | 74 | ||
| XM_021238651.1 | XP_021094310.1 | 17 - 18 | 1885 | 74 | ||
More Information
Set Membership: |
|

Map3k20
Genomic Map