Assay Details
| Gene Symbol: Map3k20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857686.3 | XP_004857743.1 | 1 - 2 | 155 | 140 | |
| XM_021238653.1 | XP_021094312.1 | 1 - 2 | 155 | 140 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004857686.3, XM_021238653.1 transcripts.
Set Membership: |
|

Map3k20
Genomic Map