Assay Details
| Gene Symbol: Serpinb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858451.2 | XP_004858508.1 | 7 - 8 | 815 | 83 | |
| XM_021239283.1 | XP_021094942.1 | 7 - 8 | 890 | 83 | ||
| XM_021239284.1 | XP_021094943.1 | 6 - 7 | 708 | 83 | ||
| XM_021239285.1 | XP_021094944.1 | 5 - 6 | 1053 | 83 | ||
More Information
Set Membership: |
|

Serpinb13
Genomic Map