Assay Details
| Gene Symbol: LOC101707650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859395.2 | XP_004859452.1 | 7 - 8 | 1379 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707650| Gene Symbol: LOC101707650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859395.2 | XP_004859452.1 | 7 - 8 | 1379 | 82 | |
Set Membership: |
|