Assay Details
| Gene Symbol: Skap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859601.2 | XP_004859658.1 | 5 - 6 | 441 | 62 | |
| XM_013077779.2 | XP_012933233.1 | 5 - 6 | 441 | 62 | ||
| XM_013077781.2 | XP_012933235.1 | 5 - 6 | 441 | 62 | ||
| XM_021240062.1 | XP_021095721.1 | 5 - 6 | 503 | 62 | ||
More Information
Set Membership: |
|

Skap1
Genomic Map