Assay Details
| Gene Symbol: CUNH17orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861039.3 | XP_004861096.1 | 4 - 5 | 601 | 62 | |
| XM_004861041.3 | XP_004861098.1 | 4 - 5 | 414 | 62 | ||
| XM_021241291.1 | XP_021096950.1 | 4 - 5 | 418 | 62 | ||
More Information
Set Membership: |
|

CUNH17orf62
Genomic Map