Assay Details
| Gene Symbol: LOC101702503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864212.2 | XP_004864269.1 | 3 - 4 | 346 | 71 | |
| XM_021243911.1 | XP_021099570.1 | 3 - 4 | 340 | 71 | ||
More Information
Set Membership: |
|

LOC101702503
Genomic Map