Assay Details
| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871287.3 | XP_004871344.1 | 4 - 5 | 502 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtcap2| Gene Symbol: Krtcap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871287.3 | XP_004871344.1 | 4 - 5 | 502 | 59 | |
Set Membership: |
|