Assay Details
| Gene Symbol: Lgals12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069081.2 | XP_012924535.2 | 6 - 7 | 679 | 69 | |
| XM_021251865.1 | XP_021107524.1 | 6 - 7 | 698 | 69 | ||
| XM_021251866.1 | XP_021107525.1 | 5 - 6 | 578 | 69 | ||
| XM_021251867.1 | XP_021107526.1 | 6 - 7 | 792 | 69 | ||
| XM_021251868.1 | XP_021107527.1 | 6 - 7 | 745 | 69 | ||
| XM_021251869.1 | XP_021107528.1 | 7 - 8 | 815 | 69 | ||
| XM_021251870.1 | XP_021107529.1 | 7 - 8 | 924 | 69 | ||
More Information
Set Membership: |
|

Lgals12
Genomic Map