Assay Details
| Gene Symbol: Mvb12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073796.2 | XP_012929250.1 | 5 - 6 | 624 | 61 | |
| XM_013073797.2 | XP_012929251.1 | 5 - 6 | 622 | 61 | ||
| XM_021262046.1 | XP_021117705.1 | 5 - 6 | 593 | 61 | ||
| XM_021262047.1 | XP_021117706.1 | 5 - 6 | 626 | 61 | ||
| XM_021262048.1 | XP_021117707.1 | 5 - 6 | 624 | 61 | ||
| XM_021262049.1 | XP_021117708.1 | 5 - 6 | 624 | 61 | ||
More Information
Set Membership: |
|

Mvb12b
Genomic Map