Assay Details
| Gene Symbol: Iqca1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078021.1 | XP_012933475.1 | 7 - 8 | 1118 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqca1l| Gene Symbol: Iqca1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078021.1 | XP_012933475.1 | 7 - 8 | 1118 | 100 | |
Set Membership: |
|