Assay Details
| Gene Symbol: Rgs18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013078905.2 | XP_012934359.1 | 2 - 3 | 397 | 105 | |
| XM_021242917.1 | XP_021098576.1 | 3 - 4 | 499 | 105 | ||
More Information
Set Membership: |
|

Rgs18
Genomic Map