Assay Details
| Gene Symbol: C2cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237513.1 | XP_021093172.1 | 12 - 13 | 1252 | 85 | |
| XM_021237514.1 | XP_021093173.1 | 11 - 12 | 1207 | 85 | ||
| XM_021237515.1 | XP_021093174.1 | 13 - 14 | 1228 | 85 | ||
| XM_021237517.1 | XP_021093176.1 | 12 - 13 | 1252 | 85 | ||

C2cd6
Genomic Map