Assay Details
| Gene Symbol: C2cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237513.1 | XP_021093172.1 | 14 - 15 | 1778 | 100 | |
| XM_021237514.1 | XP_021093173.1 | 13 - 14 | 1733 | 100 | ||
| XM_021237515.1 | XP_021093174.1 | 15 - 16 | 1754 | 100 | ||
| XM_021237516.1 | XP_021093175.1 | 13 - 14 | 1487 | 100 | ||

C2cd6
Genomic Map