Assay Details
| Gene Symbol: C2cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237513.1 | XP_021093172.1 | 1 - 2 | 317 | 94 | |
| XM_021237514.1 | XP_021093173.1 | 1 - 2 | 317 | 94 | ||
| XM_021237516.1 | XP_021093175.1 | 1 - 2 | 317 | 94 | ||
| XM_021237517.1 | XP_021093176.1 | 1 - 2 | 317 | 94 | ||

C2cd6
Genomic Map