Assay Details
| Gene Symbol: C2cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237513.1 | XP_021093172.1 | 3 - 4 | 457 | 86 | |
| XM_021237514.1 | XP_021093173.1 | 3 - 4 | 457 | 86 | ||
| XM_021237515.1 | XP_021093174.1 | 4 - 5 | 433 | 86 | ||
| XM_021237516.1 | XP_021093175.1 | 3 - 4 | 457 | 86 | ||
| XM_021237517.1 | XP_021093176.1 | 3 - 4 | 457 | 86 | ||

C2cd6
Genomic Map