Assay Details
| Gene Symbol: C2cd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237513.1 | XP_021093172.1 | 7 - 8 | 841 | 87 | |
| XM_021237514.1 | XP_021093173.1 | 6 - 7 | 796 | 87 | ||
| XM_021237515.1 | XP_021093174.1 | 8 - 9 | 817 | 87 | ||
| XM_021237516.1 | XP_021093175.1 | 7 - 8 | 841 | 87 | ||
| XM_021237517.1 | XP_021093176.1 | 7 - 8 | 841 | 87 | ||

C2cd6
Genomic Map