Assay Details
| Gene Symbol: Tmem121b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249644.1 | XP_021105303.1 | 1 | 2624 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem121b| Gene Symbol: Tmem121b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249644.1 | XP_021105303.1 | 1 | 2624 | 109 | |
Set Membership: |
|