Assay Details
Gene Symbol: Luzp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021252179.1 | XP_021107838.1 | 2 - 3 | 299 | 97 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Luzp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021252179.1 | XP_021107838.1 | 2 - 3 | 299 | 97 |