Assay Details
| Gene Symbol: CUNH5orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261487.1 | XP_021117146.1 | 28 - 29 | 5880 | 119 | |
| XM_021261488.1 | XP_021117147.1 | 28 - 29 | 5880 | 119 | ||
| XM_021261489.1 | XP_021117148.1 | 28 - 29 | 5880 | 119 | ||
| XM_021261490.1 | XP_021117149.1 | 28 - 29 | 5880 | 119 | ||
| XM_021261491.1 | XP_021117150.1 | 28 - 29 | 5880 | 119 | ||
More Information
Set Membership: |
|

CUNH5orf42
Genomic Map