Assay Details
| Gene Symbol: Hyal3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834101.3 | XP_004834158.1 | 2 - 3 | 138 | 96 | |
| XM_004834102.3 | XP_004834159.1 | 2 - 3 | 138 | 96 | ||
More Information
Set Membership: |
|

Hyal3
Genomic Map