Assay Details
| Gene Symbol: Hrh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841001.2 | XP_004841058.1 | 1 - 2 | 257 | 70 | |
| XM_021255209.1 | XP_021110868.1 | 1 - 2 | 257 | 70 | ||
| XM_021255210.1 | XP_021110869.1 | 1 - 2 | 257 | 70 | ||
More Information
Set Membership: |
|

Hrh3
Genomic Map