Assay Details
| Gene Symbol: Adm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845313.2 | XP_004845370.1 | 2 - 3 | 292 | 101 | |
| XM_021258714.1 | XP_021114373.1 | 3 - 4 | 666 | 101 | ||
More Information
Set Membership: |
|

Adm2
Genomic Map