Assay Details
| Gene Symbol: Mapk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845327.3 | XP_004845384.1 | 3 - 4 | 315 | 61 | |
| XM_021258735.1 | XP_021114394.1 | 4 - 5 | 387 | 61 | ||
| XM_021258736.1 | XP_021114395.1 | 4 - 5 | 387 | 61 | ||
| XM_021258737.1 | XP_021114396.1 | 3 - 4 | 315 | 61 | ||
| XM_021258738.1 | XP_021114397.1 | 5 - 6 | 460 | 61 | ||
| XM_021258739.1 | XP_021114398.1 | 4 - 5 | 445 | 61 | ||
| XM_021258740.1 | XP_021114399.1 | 4 - 5 | 387 | 61 | ||
| XM_021258741.1 | XP_021114400.1 | 4 - 5 | 387 | 61 | ||
More Information
Set Membership: |
|

Mapk11
Genomic Map