Assay Details
| Gene Symbol: Slc31a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849559.3 | XP_004849616.1 | 4 - 5 | 610 | 69 | |
| XM_013073634.2 | XP_012929088.1 | 4 - 5 | 580 | 69 | ||
More Information
Set Membership: |
|

Slc31a1
Genomic Map