Assay Details
| Gene Symbol: Tmem178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851291.2 | XP_004851348.1 | 2 - 3 | 496 | 78 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem178b| Gene Symbol: Tmem178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851291.2 | XP_004851348.1 | 2 - 3 | 496 | 78 | |