Assay Details
| Gene Symbol: Zxdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854146.3 | XP_004854203.1 | 5 - 6 | 1706 | 54 | |
| XM_004854147.3 | XP_004854204.1 | 5 - 6 | 1701 | 54 | ||
| XM_021265993.1 | XP_021121652.1 | 5 - 6 | 1705 | 54 | ||
More Information
Set Membership: |
|

Zxdc
Genomic Map