Assay Details
| Gene Symbol: Lipf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858363.3 | XP_004858420.1 | 8 - 9 | 962 | 68 | |
| XM_021239168.1 | XP_021094827.1 | 8 - 9 | 968 | 68 | ||
| XM_021239169.1 | XP_021094828.1 | 8 - 9 | 863 | 68 | ||
More Information
Set Membership: |
|

Lipf
Genomic Map