Assay Details
Gene Symbol: Fam71e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866819.3 | XP_004866876.1 | 1 - 2 | 209 | 72 | |
XM_004866820.3 | XP_004866877.1 | 1 - 2 | 209 | 72 | ||
XM_004866821.3 | XP_004866878.1 | 1 - 2 | 209 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004866819.3, XM_004866820.3, XM_004866821.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
